Potassium chloride 99-100,5 Argentometric titration, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC, E508 Al_0,0001, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Potassiumchloride

12636-6X1KG 12636-500G 12636-5KG
BJ12636-1KGCS 577.72 USD
BJ12636-1KG BJ12636-250G BJ12636-500G BJ-12636-2.5KG BJ-12636-5KG
Potassium chloride 99-100,5 Argentometric titration, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC, E508 Al_0,0001, Fluka™
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (105°C, 3h) Max. 0.5 %
Acidity or alkalinity complying
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.5 ppm
Barium (Ba) complying
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 1 ppm
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Magnesium (Mg) Max. 0.005 %
Manganese (Mn) complying
Sodium (Na) Max. 0.07 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 5 ppm
Mg, earth alkali metals (as Ca) Max. 200 ppm
Bromide (Br) Max. 0.1 %
Iodide (I) test complying
Iodide (I) Max. 0.005 %
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR