Bis(pentamethylcyclopentadienyl)magnesium ≥99.999%, PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger

97-1045-1G 97-1045-5G 97-1045-25G
100201-292EA 314.22 USD
100201-292 100197-036 100199-306
Bis(pentamethylcyclopentadienyl)magnesium ≥99.999%, PURATREM, Electronic Grade
Bis(pentamethylcyclopentadienyl)magnesium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₀H₃₀Mg
MW: 294.757 g/mol
MDL Number: MFCD00151387
CAS Number: 74507-64-5
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR