Bis(cyclopentadienyl)magnesium ≥99.99%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
97-1040-25G
97-1040-1G
97-1040-5G
100210-600EA
1943.32
USD
100210-600
100200-224
100200-226
Bis(cyclopentadienyl)magnesium ≥99.99%, PURATREM
Bis(cyclopentadienyl)magnesium
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₁₀H₁₀Mg MW: 154.49 g/mol Boiling Pt: 290 °C/ 160 °C (0.1 mmHg) (dec./ subl.) Melting Pt: 176 °C |
MDL Number:
MFCD00134487 CAS Number: 1284-72-6 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...