Bismuth(III) titanate ≥98%

Supplier: STREM CHEMICALS INC MS

93-8312-100G 93-8312-25G
100210-588EA 123.14 USD
100210-588 100205-796
Bismuth(III) titanate ≥98%
Bismuth(III) titanate
MDL Number: MFCD00136014
CAS Number: 12048-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR