Lead(II) hexafluoroacetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS

93-8265-1G 93-8265-5G
100204-448EA 103.32 USD
100204-448 100202-326
Lead(II) hexafluoroacetylacetonate ≥98%
Lead(II) hexafluoroacetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Boiling Pt: 210 °C/ 180 °C (0,05 mmHg) (dec./ subl.)
Melting Pt: 153…158 °C
MDL Number: MFCD00058833
CAS Number: 19648-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR