Lead(II) zirconate ≥99%

Supplier: STREM CHEMICALS INC MS

93-8239-500G 93-8239-100G
100203-876EA 1272.43 USD
100203-876 100199-670
Lead(II) zirconate ≥99%
Lead(II) zirconate
Formula: O₃PbZr
MW: 346.421 g/mol
Density: 7,0
MDL Number: MFCD00016275
CAS Number: 12060-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR