Lead(II)-2-ethylhexanoate (40,5 - 42,5% Pb)

Supplier: STREM CHEMICALS INC MS

Synonyms: Lead(II) 2-ethylhexanoate

93-8224-1000G 93-8224-250G
100202-320EA 142.95 USD
100202-320 100202-322
Lead(II)-2-ethylhexanoate (40,5 - 42,5% Pb)
Lead(II)-2-ethylhexanoate
Formula: C₁₆H₃₀O₄Pb
MW: 493.61 g/mol
Density: 1,56
Flash Pt: 324 °F
MDL Number: MFCD00014003
CAS Number: 301-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR