Lead antimonide ≥99%

Supplier: STREM CHEMICALS INC MS

93-8201-5G 93-8201-25G
100210-578EA 124.55 USD
100210-578 100202-312
Lead antimonide ≥99%
Lead antimonide
Formula: PbSb
MW: 328.96 g/mol
MDL Number: MFCD00151190
CAS Number: 12266-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR