Thallium(III) acetate hydrate

Supplier: STREM CHEMICALS INC MS
Danger

93-8101-50G 93-8101-10G
100200-610EA 1886.71 USD
100200-610 100197-034
Thallium(III) acetate hydrate
Thallium(III) acetate hydrate
MDL Number: MFCD00149571
CAS Number: 62811-75-0
UN: 1707
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR