(1,5-Cyclooctadiene)diiodoplatinum(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diiodo(1,5-cyclooctadiene)platinum(II)

93-7819-1G 93-7819-5G
100208-736EA 505.29 USD
100208-736 100205-904
(1,5-Cyclooctadiene)diiodoplatinum(II) ≥99%
(1,5-Cyclooctadiene)diiodoplatinum(II)
Formula: C₈H₁₂I₂Pt
MW: 557.077 g/mol
MDL Number: MFCD00050491
CAS Number: 12266-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR