Dibromo(1,5-cyclooctadiene)platinum(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-7816-1G 93-7816-5G
100210-906EA 208.06 USD
100210-906 100200-208
Dibromo(1,5-cyclooctadiene)platinum(II) ≥98%
Dibromo(1,5-cyclooctadiene)platinum(II)
Formula: C₈H₁₂Br₂Pt
MDL Number: MFCD00058724
CAS Number: 12145-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR