Iridium(IV) chloride hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-7727-5G 93-7727-1G
100208-598EA 1670.15 USD
100208-598 100196-826
Iridium(IV) chloride hydrate
Iridium(IV) chloride hydrate
Formula: Cl₄Ir·xH₂O
Melting Pt: Dec.
MDL Number: MFCD00150243
CAS Number: 207399-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR