Potassium osmate(VI) dihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate
93-7613-1G
93-7613-250MG
93-7613-5G
100207-164EA
264.68
USD
100207-164
100207-162
100207-166
Potassium osmate(VI) dihydrate ≥99%
Potassium osmate(VI) dihydrate
Formula:
K₂O₄Os·2H₂O MW: 368.45 g/mol |
MDL Number:
MFCD00149919 CAS Number: 10022-66-9 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...