Ammonium metatungstate hydrate ≥99.9% (W basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium meta-tungsten oxide hydrate
93-7420-100G
93-7420-25G
100200-200EA
147.2
USD
100200-200
100207-156
Ammonium metatungstate hydrate ≥99.9% (W basis)
Ammonium metatungstate hydrate
Formula:
(NH₄)₆W₁₂O₃₉·xH₂O MW: 2938.41 g/mol (anhy) |
MDL Number:
MFCD00042673 CAS Number: 12028-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...