Tantalum(V) methanolate ≥99.99% (Ta basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tantalum(V) methoxide

93-7329-2G 93-7329-10G
100205-756EA 162.77 USD
100205-756 100205-754
Tantalum(V) methanolate ≥99.99% (Ta basis), PURATREM
Tantalum(V) methanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₅H₁₅O₅Ta
MW: 336.12 g/mol
Boiling Pt: 189 °C (10 mmHg)
Melting Pt: 50 °C
MDL Number: MFCD00058817
CAS Number: 865-35-0
UN: 3178
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR