Lutetium(III) perchlorate 50% in aqueous solution ≥99.9% (Lu basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-7118-10G 93-7118-2G
100209-210EA 726.09 USD
100209-210 100197-954
Lutetium(III) perchlorate 50% in aqueous solution ≥99.9% (Lu basis)
Lutetium (III) perchlorate
Formula: Cl₃LuO₁₂
MW: 473.319 g/mol
MDL Number: MFCD00016190
CAS Number: 14646-29-8
UN: 3211
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR