Lutetium(III) sulfate hydrate ≥99.9% (Lu basis)

Supplier: STREM CHEMICALS INC MS

93-7116-5G 93-7116-1G
100210-896EA 452.92 USD
100210-896 100200-796
Lutetium(III) sulfate hydrate ≥99.9% (Lu basis)
Lutetium(III) sulfate hydrate
Formula: Lu₂O₁₂S₃·xH₂O
MW: 683.12 g/mol (anhy)
MDL Number: MFCD00149953
CAS Number: 13473-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR