Lutetium(III) acetate hydrate ≥99.9% (Lu basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-7110-5G 93-7110-1G
100199-176EA 373.66 USD
100199-176 100199-174
Lutetium(III) acetate hydrate ≥99.9% (Lu basis)
Lutetium(III) acetate hydrate
Formula: C₆H₉LuO₆·xH₂O
MW: 352.11 g/mol (anhy)
MDL Number: MFCD00150119
CAS Number: 207500-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR