Ytterbium (III) sulfate octahydrate ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Ytterbium(III) sulphate octahydrate
93-7027-5G
93-7027-25G
100199-170EA
48.12
USD
100199-170
100202-274
Ytterbium (III) sulfate octahydrate ≥99.9% (Yb basis)
Ytterbium (III) sulphate octahydrate
Formula:
O₁₂S₃Yb₂·8H₂O MW: 778.396 g/mol Density: 3.286 |
MDL Number:
MFCD00149887 CAS Number: 10034-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...