Ytterbium triacetate hydrate ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ytterbium(III) acetate hydrate
93-7001-50G
93-7001-10G
100205-902EA
230.71
USD
100205-902
100197-946
Ytterbium triacetate hydrate ≥99.9% (Yb basis)
Ytterbium triacetate hydrate
Formula:
C₆H₉O₆Yb·H₂O MW: 368.19 g/mol Density: 2,09 |
MDL Number:
MFCD00150127 CAS Number: 15280-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...