Thulium(III) oxalate hydrate ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS

93-6912-1G 93-6912-5G
100205-742EA 77.85 USD
100205-742 100203-572
Thulium(III) oxalate hydrate ≥99.9% (Tm basis)
Thulium(III) oxalate hydrate
Formula: C₆O₁₂Tm₂·xH₂O
MW: 601.93 g/mol (anhy)
MDL Number: MFCD00050804
CAS Number: 58176-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR