Thulium(III) acetate tetrahydrate ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6911-5G 93-6911-1G
100208-228EA 271.75 USD
100208-228 100203-570
Thulium(III) acetate tetrahydrate ≥99.9% (Tm basis)
Thulium(III) acetate tetrahydrate
MDL Number: MFCD00150125
CAS Number: 34431-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR