Thulium(III) 2,4-pentanedionate hydrate ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Thulium(III) acetylacetonate hydrate, Thulium(III) acetylacetonate trihydrate

93-6910-5G 93-6910-1G
100207-900EA 1548.43 USD
100207-900 100207-898
Thulium(III) 2,4-pentanedionate hydrate ≥99.9% (Tm basis)
Thulium(III) 2,4-pentanedionate hydrate
Formula: C₁₅H₂₁O₆Tm·xH₂O
MW: 466.26 g/mol (anhy)
MDL Number: MFCD00150646
CAS Number: 14589-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR