Thulium(III) chloride hydrate ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS

93-6902-5G 93-6902-1G
100203-568EA 273.17 USD
100203-568 100209-198
Thulium(III) chloride hydrate ≥99.9% (Tm basis)
Thulium(III) chloride hydrate
Formula: Cl₃Tm·H₂O
MW: 293.309 g/mol
Melting Pt: 821 °C (anhydr.)
MDL Number: MFCD00149877
CAS Number: 19423-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR