Erbium triacetate tetrahydrate ≥99.9% (Er basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Erbium(III) acetate tetrahydrate
93-6824-50G
93-6824-10G
100196-816EA
226.46
USD
100196-816
100210-536
Erbium triacetate tetrahydrate ≥99.9% (Er basis)
Erbium triacetate tetrahydrate
Formula:
C₆H₉ErO₆·4H₂O MW: 416.45 g/mol Density: 2.114 |
MDL Number:
MFCD00013034 CAS Number: 15280-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...