Erbium(III) carbonate hydrate ≥99.9% (Er basis)
Supplier: STREM CHEMICALS INC MS
93-6822-50G
93-6822-10G
100202-828EA
484.06
USD
100202-828
100205-738
Erbium(III) carbonate hydrate ≥99.9% (Er basis)
Erbium(III) carbonate hydrate
Formula:
Er₂(CO₃)₃.xH₂O MW: 514.545 g/mol (anhy) |
MDL Number:
MFCD00149951 CAS Number: 22992-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...