Erbium(III) perchlorate 6H2O 50% in aqueous solution ≥99.9% (Er basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-6821-2G 93-6821-10G
100197-942EA 36.8 USD
100197-942 100202-718
Erbium(III) perchlorate 6H2O 50% in aqueous solution ≥99.9% (Er basis)
Erbium(III) perchlorate 6H2O
MDL Number: MFCD00016073
CAS Number: 61565-07-9
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR