Erbium(III)-2,4-pentanedionate hydrate ≥99.9% (Er basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Erbium(III) acetylacetonate hydrate, Erbium acetylacetonate hydrate
93-6801-5G
93-6801-25G
100199-652EA
157.11
USD
100199-652
100207-490
Erbium(III)-2,4-pentanedionate hydrate ≥99.9% (Er basis)
Erbium(III)-2,4-pentanedionate hydrate
Formula:
C₁₅H₂₁ErO₆·xH₂O MW: 464.59 g/mol (anhy) |
MDL Number:
MFCD00150165 CAS Number: 14553-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...