Dysprosium(III) isopropoxide ≥99.9% (Dy basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-6619-5G 93-6619-1G
100202-826EA 314.22 USD
100202-826 100205-734
Dysprosium(III) isopropoxide ≥99.9% (Dy basis)
Dysprosium (III) isopropoxide

Alkoxide

Formula: C₉H₂₁DyO₃
MDL Number: MFCD00049784
CAS Number: 6742-68-3
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR