Dysprosium(III) perchlorate hexahydrate 50% in aqueous solution ≥99.9% (dye basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Dysprosium(III) perchlorate
93-6617-10G
93-6617-50G
100197-936EA
128.8
USD
100197-936
100209-192
Dysprosium(III) perchlorate hexahydrate 50% in aqueous solution ≥99.9% (dye basis)
Dysprosium (III) perchlorate hexahydrate
Formula:
Cl₃DyO₁₂·6H₂O MW: 568.94 g/mol |
MDL Number:
MFCD00150227 CAS Number: 14692-17-2 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...