Dysprosium(III) acetate tetrahydrate ≥99.9% (dye basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6601-25G 93-6601-100G
100205-730EA 75.02 USD
100205-730 100198-434
Dysprosium(III) acetate tetrahydrate ≥99.9% (dye basis)
Dysprosium (III) acetate tetrahydrate
Formula: C₆H₉DyO₆·4H₂O
MW: 411.68 g/mol
Melting Pt: 120 °C (dec.)
MDL Number: MFCD00013033
CAS Number: 15280-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR