Terbium(III) perchlorate 50% in aqueous solution ≥99.9% (Tb basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-6512-10G 93-6512-2G
100196-810EA 232.12 USD
100196-810 100210-520
Terbium(III) perchlorate 50% in aqueous solution ≥99.9% (Tb basis)
Terbium (III) perchlorate
Formula: Cl₃O₁₂Tb
MDL Number: MFCD00150362
CAS Number: 14014-09-6
UN: 3211
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR