Terbium(III) acetylacetonate hydrate ≥99.9% (Tb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Terbium(III) acetylacetonate trihydrate, Terbium(III) 2,4-pentanedionate hydrate
93-6501-1G
93-6501-5G
100210-518EA
56.62
USD
100210-518
100196-804
Terbium(III) acetylacetonate hydrate ≥99.9% (Tb basis)
Terbium(III) acetylacetonate hydrate
Formula:
C₁₅H₂₁O₆Tb·xH₂O MW: 456.25 g/mol (anhy) Melting Pt: 168…170 °C |
MDL Number:
MFCD00013503 CAS Number: 14284-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...