Gadolinium(III) acetate tetrahydrate ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6424-100G 93-6424-25G
100205-900EA 243.45 USD
100205-900 100205-728
Gadolinium(III) acetate tetrahydrate ≥99.9% (Gd basis)
Gadolinium(III) acetate tetrahydrate
Density: 1,611
MDL Number: MFCD00150117
CAS Number: 15280-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR