Gadolinium trichloride hydrate ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Gadolinium(III) chloride hydrate

93-6421-25G 93-6421-100G
100205-724EA 82.09 USD
100205-724 100210-516
Gadolinium trichloride hydrate ≥99.9% (Gd basis)
Gadolinium trichloride hydrate
Formula: Cl₃Gd·H₂O
MW: 281.624 g/mol
Density: 2,424
MDL Number: MFCD00149732
CAS Number: 19423-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR