Gadolinium(III) perchlorate 40 - 50% in aqueous solution ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS

93-6420-50G 93-6420-10G
100200-784EA 353.85 USD
100200-784 100209-178
Gadolinium(III) perchlorate 40 - 50% in aqueous solution ≥99.9% (Gd basis)
Gadolinium (III) perchlorate
Formula: Cl₃GdO₁₂
MDL Number: MFCD00150239
CAS Number: 14017-52-8
UN: 3211
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR