Gadolinium(III)-2,4-pentanedionate hydrate ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6401-5G 93-6401-25G
100205-722EA 69.35 USD
100205-722 100202-822
Gadolinium(III)-2,4-pentanedionate hydrate ≥99.9% (Gd basis)
Gadolinium(III)-2,4-pentanedionate hydrate
Formula: C₁₅H₂₁GdO₆·xH₂O
MW: 454.58 g/mol (anhy)
MDL Number: MFCD00013493
CAS Number: 64438-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR