Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)europium(III) hydrate ≥99.9% (Eu basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Eu(fod)3 hydrate, Resolve-Al™ EuFOD, Europium(III)-tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate) hydrate, Europium-fod, Siever's reagent
93-6326-1G
93-6326-5G
100200-186EA
69.35
USD
100200-186
100207-130
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)europium(III) hydrate ≥99.9% (Eu basis)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)europium(III) hydrate
Formula:
C₃₀H₃₀EuF₂₁O₆·xH₂O MW: 1037.49 g/mol (anhy) |
MDL Number:
MFCD01074933 CAS Number: 17631-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...