Samarium(III) acetylacetonate hydrate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6226-100G 93-6226-25G
100203-542EA 1198.83 USD
100203-542 100204-402
Samarium(III) acetylacetonate hydrate ≥99.9% (Sm basis)
Samarium(III) acetylacetonate hydrate
MDL Number: MFCD00792904
CAS Number: 14589-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR