Samarium(III) trifluoroacetylacetonate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6219-5G 93-6219-1G
100202-246EA 397.72 USD
100202-246 100204-400
Samarium(III) trifluoroacetylacetonate ≥99.9% (Sm basis)
Samarium(III) trifluoroacetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

MDL Number: MFCD00058837
CAS Number: 23301-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR