Samarium (III) sulfate octahydrate ≥99.9% (Sm basis)
Supplier: STREM CHEMICALS INC MS
93-6216-100G
93-6216-25G
100199-292EA
376.49
USD
100199-292
100200-782
Samarium (III) sulfate octahydrate ≥99.9% (Sm basis)
Samarium (III) sulphate octahydrate
Formula:
O₁₂S₃Sm₂·8H₂O MW: 733.03 g/mol Density: 2,930 |
MDL Number:
MFCD00011236 CAS Number: 13465-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...