Samarium(III) naphthenate (8 - 10% Sm)

Supplier: STREM CHEMICALS INC MS

Synonyms: Rare-earth naphthenate, Naphthenic acids, rareearth salts

93-6206-25G 93-6206-100G
100205-710EA 757.23 USD
100205-710 100206-050
Samarium(III) naphthenate (8 - 10% Sm)
Samarium(III) naphthenate
Formula: 3(C₁₁H₇O₂).Sm
MDL Number: MFCD00148244
CAS Number: 61790-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR