Neodymium(III) oxalate decahydrate ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS

93-6034-50G 93-6034-10G
100202-240EA 390.65 USD
100202-240 100204-396
Neodymium(III) oxalate decahydrate ≥99.9% (Nd basis)
Neodymium(III) oxalate decahydrate
MDL Number: MFCD00150486
CAS Number: 14551-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR