Neodymium(III)-1,1,1-trifluoro-2,4-pentanedionate ≥99.9% (Nd basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6017-1G
93-6017-5G
100196-788EA
67.94
USD
100196-788
100210-512
Neodymium(III)-1,1,1-trifluoro-2,4-pentanedionate ≥99.9% (Nd basis)
Neodymium(III)-1,1,1-trifluoro-2,4-pentanedionate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₅H₁₂F₉NdO₆ Melting Pt: 140…142 °C |
MDL Number:
MFCD00058831 CAS Number: 37473-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...