Neodymium(III) perchlorate 50% in aqueous solution ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Perchloric acid neodymium salt

93-6010-50G 93-6010-10G
100208-212EA 426.03 USD
100208-212 100196-786
Neodymium(III) perchlorate 50% in aqueous solution ≥99.9% (Nd basis)
Neodymium(III) perchlorate
Formula: Nd(ClO₄)₃
MW: 442.59 g/mol
MDL Number: MFCD00016244
CAS Number: 13498-06-1
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR