Praseodymium ≥99.9%, Powder

Supplier: STREM CHEMICALS INC MS
Warning

93-5940-25G 93-5940-5G
100202-570EA 342.52 USD
100202-570 100196-782
Praseodymium ≥99.9%, Powder
Praseodymium
Formula: Pr
MW: 140.91 g/mol
Boiling Pt: 3127 °C
Melting Pt: 935 °C
Density: 6,782
MDL Number: MFCD00011174
CAS Number: 7440-10-0
UN: 2813
ADR: 4.3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR