Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥99% (99.9%-Pr)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III), Pr(TMHD)3

93-5937-1G 93-5937-5G
100210-510EA 52.37 USD
100210-510 100204-742
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥99% (99.9%-Pr)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Pr
MW: 690.71 g/mol
Boiling Pt: 300 °C/ 150 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 212…214 °C
MDL Number: MFCD00000027
CAS Number: 15492-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR