Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥99% (99.9%-Pr)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Pr(FOD)3, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)praseodymium(III), Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III)
93-5920-1G
93-5920-5G
100197-918EA
174.09
USD
100197-918
100209-166
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥99% (99.9%-Pr)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III)
Formula:
C₃₀H₃₀F₂₁O₆Pr MW: 1026.43 g/mol Boiling Pt: 130 °C (0.01 mmHg) (subl.) Melting Pt: 215…219 °C |
MDL Number:
MFCD00064656 CAS Number: 17978-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...