Praseodymium(III) acetate hydrate ≥99.9% (Pr basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Praseodynium acetate hydrate

93-5901-250G 93-5901-50G 93-5901-10G
100210-502EA 472.74 USD
100210-502 100210-504 100205-690
Praseodymium(III) acetate hydrate ≥99.9% (Pr basis)
Praseodymium(III) acetate hydrate
Formula: Pr(OOCCH₃)₃·xH₂O
MW: 318.03 g/mol (anhy)
MDL Number: MFCD00150121
CAS Number: 6192-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR