Cerium(III)-2-ethylhexanoate 49% in 2-ethylhexanoic acid (12% Ce)

Supplier: STREM CHEMICALS INC MS

Synonyms: Cerium(III) 2-ethylhexanoate

93-5833-2000G 93-5833-500G
100210-500EA 484.06 USD
100210-500 100205-688
Cerium(III)-2-ethylhexanoate 49% in 2-ethylhexanoic acid (12% Ce)
Cerium (III) 2-ethylhexanoate
Formula: C₂₄H₄₅CeO₆
MW: 569.727 g/mol
Density: 1.08
Flash Pt: 230 °F
MDL Number: MFCD00014004
CAS Number: 56797-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR